Learn More
4-Chlorocinnamic acid, predominantly trans, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1530414
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-Chlorocinnamic acid | |
1615-02-7 | |
C9H7ClO2 | |
25 g | |
4-chlorocinnamic acid, p-chlorocinnamic acid, 3-4-chlorophenyl acrylic acid, trans-4-chlorocinnamic acid, 2e-3-4-chlorophenyl prop-2-enoic acid, e-p-chlorocinnamic acid, cinnamic acid, p-chloro, trans-p-chlorocinnamic acid, 2-propenoic acid, 3-4-chlorophenyl, 3-p-chlorophenyl acrylic acid | |
OC(=O)\C=C\C1=CC=C(Cl)C=C1 | |
182.60 | |
CHEBI:61116 | |
99% |
predominantly trans | |
248°C to 253°C | |
MFCD00004396 | |
1365308 | |
GXLIFJYFGMHYDY-ZZXKWVIFSA-N | |
(E)-3-(4-chlorophenyl)prop-2-enoic acid | |
637797 | |
182.61 |
Chemical Identifiers
1615-02-7 | |
182.60 | |
GXLIFJYFGMHYDY-ZZXKWVIFSA-N | |
637797 | |
(E)-3-(4-chlorophenyl)prop-2-enoic acid |
C9H7ClO2 | |
MFCD00004396 | |
4-chlorocinnamic acid, p-chlorocinnamic acid, 3-4-chlorophenyl acrylic acid, trans-4-chlorocinnamic acid, 2e-3-4-chlorophenyl prop-2-enoic acid, e-p-chlorocinnamic acid, cinnamic acid, p-chloro, trans-p-chlorocinnamic acid, 2-propenoic acid, 3-4-chlorophenyl, 3-p-chlorophenyl acrylic acid | |
CHEBI:61116 | |
OC(=O)\C=C\C1=CC=C(Cl)C=C1 |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
EINECSNumber : 216-564-8
RTECSNumber : GD8490000
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only