Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-Chlorochalcone, 98+%

$41.26 - $122.62

Chemical Identifiers

Chemical Name or Material 4'-Chlorochalcone
CAS 956-02-5
Molecular Formula C15H11ClO
Formula Weight 242.71g/mol
MDL Number MFCD00016343
Beilstein 642281
Synonym 2-Benzylidene-4'-chloroacetophenone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1377206

 
alfa aesar™
A13772-06
5g Each for $41.26

AAA1377214

 
alfa aesar™
A13772-14
25g Each for $122.62
Description & Specifications

Specifications

Melting Point 97° to 100°C
Quantity 5g
Chemical Name or Material 4'-Chlorochalcone
CAS 956-02-5
Assay ≥98%
Molecular Formula C15H11ClO
Formula Weight 242.71g/mol
MDL Number MFCD00016343
Beilstein 642281
Synonym 2-Benzylidene-4'-chloroacetophenone
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number UD5572990
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.