Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chlorochalcone, 98+%

$51.54 - $706.68

Chemical Identifiers

Chemical Name or Material 4-Chlorochalcone
CAS 956-04-7
Molecular Formula C15H11ClO
Formula Weight 242.71
MDL Number MFCD00016345
Beilstein 1105953
Synonym 2-(4-Chlorobenzylidene)acetophenone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1597809

  SDS
alfa aesar™
A1597809
10g Each for $51.54

AAA1597830

  SDS
alfa aesar™
A1597830
250g Each for $706.68
Description & Specifications

Specifications

Formula Weight 242.71
Assay ≥98%
Melting Point 114° to 116°C
Quantity 10g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Chemical Name or Material 4-Chlorochalcone
Molecular Formula C15H11ClO
Health Hazard 1 Warning
MDL Number MFCD00016345
Beilstein 1105953
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 956-04-7
Synonym 2-(4-Chlorobenzylidene)acetophenone
RTECS Number UD5572994
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.