Please login to your online account to display your discounted pricing

Alfa Aesar™ 4'-Chlorobutyrophenone, 97%

$36.03 - $99.28

Chemical Identifiers

Chemical Name or Material 4'-Chlorobutyrophenone
CAS 4981-63-9
Molecular Formula C10H11ClO
Formula Weight 182.65
MDL Number MFCD00013647
Beilstein 1864288
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1453106

 
alfa aesar™
A1453106
5g Each for $36.03

AAA1453114

 
alfa aesar™
A1453114
25g Each for $99.28
Description & Specifications

Specifications

Formula Weight 182.65
Chemical Name or Material 4'-Chlorobutyrophenone
Assay 97%
Molecular Formula C10H11ClO
MDL Number MFCD00013647
CAS 4981-63-9
Beilstein 1864288
Boiling Point 253° to 254°C
Melting Point 35° to 37°C
Quantity 5g