missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chlorobutyronitrile, 97%

$54.46 - $54.46

Chemical Identifiers

CAS 628-20-6
Molecular Formula C4H6ClN
Linear Formula Cl(CH2)3CN
Formula Weight 103.55
MDL Number MFCD00001972
UN Number UN3276
Beilstein 1633582
Synonym 1-Chloro-3-cyanopropane
Chemical Name or Material 4-Chlorobutyronitrile
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1894218 SDS Alfa Aesar™
A1894218
50g Each for $54.46
AAA189420B SDS Alfa Aesar™
A189420B
1000g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AAA1894230 SDS Alfa Aesar™
A1894230
250g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

628-20-6
Cl(CH2)3CN
MFCD00001972
1633582
4-Chlorobutyronitrile
C4H6ClN
103.55
UN3276
1-Chloro-3-cyanopropane

Specifications

85°C (185°F)
50g
C4H6ClN
103.55
UN3276
1-Chloro-3-cyanopropane
97%
195° to 197°C
628-20-6
Cl(CH2)3CN
MFCD00001972
1633582
4-Chlorobutyronitrile

Safety and Handling

GHS H Statement
H301-H227-H315-H319-H335
Toxic if swallowed.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P301+P310-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.

RTECSNumber : ET9020000

TSCA : TSCA