Please login to your online account to display your discounted pricing

4-chlorobutyl chloroformate, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Chlorobutyl chloroformate
Name Note 97%
CAS 37693-18-8
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C5H8Cl2O2
Linear Formula ClCO2(CH2)4Cl
Formula Weight 171.02
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC314490250

  SDS
thermo fisher scientific
AC314490250
Glass bottle 25mL CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Liquid
Formula Weight 171.02
Quantity 25mL
Color Clear colorless
Density 1.2500g/mL
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 4-Chlorobutyl chloroformate
Packaging Glass bottle
Refractive Index 1.4520 to 1.4540 (20°C, 589nm)
Name Note 97%
Assay Percent Range 97%
Linear Formula ClCO2(CH2)4Cl
Health Hazard 1 GHS Signal Word: Danger
Infrared Spectrum Authentic
Molecular Formula C5H8Cl2O2
MDL Number MFCD00134472
CAS 37693-18-8
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if inhaled.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Recommended Storage Refrigerator +4°C

Coupling reagent

Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye damage.
Causes severe skin burns and eye damage.
Harmful if inhaled.

Refrigerator +4°C