Learn More
4-chlorobenzylamine, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 108790500
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-chlorobenzylamine | |
97.5 | |
Yellow | |
215.0°C | |
Authentic | |
Glass bottle | |
1.5576 to 1.5610 | |
MFCD00008121 | |
12, 1074 | |
4-chlorobenzylamine, 4-chlorophenyl methanamine, p-chlorobenzylamine, benzenemethanamine, 4-chloro, 1-4-chlorophenyl methanamine, 4-chloro-benzylamine, benzylamine, p-chloro, 4-chlorophenyl methylamine, para-chlorobenzylamine, chembl13218 | |
YMVFJGSXZNNUDW-UHFFFAOYSA-N | |
(4-chlorophenyl)methanamine | |
66036 | |
98% |
104-86-9 | |
100.0 | |
1.1600g/mL | |
90°C | |
97.5% min. (GC) | |
C7H8ClN | |
ClC6H4CH2NH2 | |
50 g | |
1.16 | |
Solubility in water: immiscible with water | |
C1=CC(=CC=C1CN)Cl | |
141.6 | |
141.6 | |
Liquid |
Chemical Identifiers
104-86-9 | |
141.6 | |
YMVFJGSXZNNUDW-UHFFFAOYSA-N | |
66036 | |
C1=CC(=CC=C1CN)Cl |
C7H8ClN | |
MFCD00008121 | |
4-chlorobenzylamine, 4-chlorophenyl methanamine, p-chlorobenzylamine, benzenemethanamine, 4-chloro, 1-4-chlorophenyl methanamine, 4-chloro-benzylamine, benzylamine, p-chloro, 4-chlorophenyl methylamine, para-chlorobenzylamine, chembl13218 | |
(4-chlorophenyl)methanamine |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specifica
GHS Signal Word: Danger
EINECSNumber : 203-245-3
RUO – Research Use Only