missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Chlorobenzyl Mercaptan, TCI America™

$84.03 - $731.83

Chemical Identifiers

CAS 6258-66-8
Molecular Formula C7H7ClS
Molecular Weight (g/mol) 158.643
MDL Number MFCD00004870
InChI Key GKQXPTHQTXCXEV-UHFFFAOYSA-N
Synonym 4-chlorobenzyl mercaptan, 4-chlorobenzenemethanethiol, 4-chlorobenzylmercaptan, 4-chlorophenyl methanethiol, p-chlorobenzyl mercaptan, benzenemethanethiol, 4-chloro, p-chlorotoluene-alpha-thiol, 4-chloro benzyl mercaptan, 4-chlor-benzylthiol, 4-chlorobenzyl thiol
PubChem CID 80409
IUPAC Name (4-chlorophenyl)methanethiol
SMILES C1=CC(=CC=C1CS)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C065825G View Documents TCI America
C065825G
25g Each for $84.03
 
C0658500G View Documents TCI America
C0658500G
500g Each for $731.83
 
Specifications

Chemical Identifiers

6258-66-8
158.643
GKQXPTHQTXCXEV-UHFFFAOYSA-N
80409
C1=CC(=CC=C1CS)Cl
C7H7ClS
MFCD00004870
4-chlorobenzyl mercaptan, 4-chlorobenzenemethanethiol, 4-chlorobenzylmercaptan, 4-chlorophenyl methanethiol, p-chlorobenzyl mercaptan, benzenemethanethiol, 4-chloro, p-chlorotoluene-alpha-thiol, 4-chloro benzyl mercaptan, 4-chlor-benzylthiol, 4-chlorobenzyl thiol
(4-chlorophenyl)methanethiol

Specifications

103°C
25g
C7H7ClS
2810
GKQXPTHQTXCXEV-UHFFFAOYSA-N
(4-chlorophenyl)methanethiol
80409
Clear Liquid at 20°C
4-Chlorobenzyl Mercaptan
Yellow
6258-66-8
MFCD00004870
4-chlorobenzyl mercaptan, 4-chlorobenzenemethanethiol, 4-chlorobenzylmercaptan, 4-chlorophenyl methanethiol, p-chlorobenzyl mercaptan, benzenemethanethiol, 4-chloro, p-chlorotoluene-alpha-thiol, 4-chloro benzyl mercaptan, 4-chlor-benzylthiol, 4-chlorobenzyl thiol
C1=CC(=CC=C1CS)Cl
158.643
158.64
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No