Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chlorobenzyl mercaptan, 98%

Chemical Identifiers

Chemical Name or Material 4-Chlorobenzyl mercaptan
CAS 6258-66-8
Molecular Formula C7H7ClS
Formula Weight 158.65
MDL Number MFCD00004870
UN Number UN2810
Beilstein 606496
Synonym 4-Chloro-alpha-toluenethiol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1987606

  SDS
alfa aesar™
A1987606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1987622

  SDS
alfa aesar™
A1987622
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1987614

  SDS
alfa aesar™
A1987614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 158.65
Chemical Name or Material 4-Chlorobenzyl mercaptan
Odor Stench
Assay 98%
Molecular Formula C7H7ClS
MDL Number MFCD00004870
CAS 6258-66-8
UN Number UN2810
Beilstein 606496
Synonym 4-Chloro-alpha-toluenethiol
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H331-H227-H302-H315-H319-H335
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P260-P280h-P305+P351+P338-P309-P310
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
Flash Point 76°C (168°F)
Boiling Point 234°C
Melting Point 19° to 20°C
Quantity 5g
Safety and Handling

Danger

GHS P Statement
P260-P280h-P305+P351+P338-P309-P310
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

GHS H Statement
H331-H227-H302-H315-H319-H335
Toxic if inhaled.
Combustible liquid.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.