missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Chlorobenzyl cyanide, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 108815000

 View more versions of this product

Catalog No. AC108815000



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

4-Chlorobenzyl cyanide
140-53-4
100.0
Colorless to Yellow
133°C
98% min. (GC)
C8H6ClN
ClC6H4CH2CN
500 g
4-chlorobenzyl cyanide, 4-chlorophenyl acetonitrile, 4-chlorophenylacetonitrile, 2-4-chlorophenyl acetonitrile, p-chlorobenzyl cyanide, 4-chlorobenzeneacetonitrile, benzeneacetonitrile, 4-chloro, p-chlorophenyl acetonitrile, p-chlorophenylacetonitrile, 4-chlorobenzylcyanide
IVYMIRMKXZAHRV-UHFFFAOYSA-N
2-(4-chlorophenyl)acetonitrile
241582
151.6
Liquid After Melting
98+%
98.0
29°C to 30°C
265°C to 267°C
Authentic
Glass bottle
1.5415 to 1.5435
MFCD00001918
09, 448
Solubility in water: insoluble in water. Other solubilities: soluble in acetone and alcohol
C1=CC(=CC=C1CC#N)Cl
151.6
CHEBI:17346
98+%

Chemical Identifiers

140-53-4
151.6
IVYMIRMKXZAHRV-UHFFFAOYSA-N
241582
2-(4-chlorophenyl)acetonitrile
C8H6ClN
MFCD00001918
4-chlorobenzyl cyanide, 4-chlorophenyl acetonitrile, 4-chlorophenylacetonitrile, 2-4-chlorophenyl acetonitrile, p-chlorobenzyl cyanide, 4-chlorobenzeneacetonitrile, benzeneacetonitrile, 4-chloro, p-chlorophenyl acetonitrile, p-chlorophenylacetonitrile, 4-chlorobenzylcyanide
CHEBI:17346
C1=CC(=CC=C1CC#N)Cl
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Toxic in contact with skin.
Toxic if inhaled.
Fatal if swallowed.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF IN EYES:

GHS Signal Word: Danger

EINECSNumber : 205-418-9

RTECSNumber : AL8250000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only