missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Chlorobenzyl cyanide, 98+%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 108812500

 View more versions of this product

Catalog No. AC108812500

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Liquid After Melting
151.6
Clear colorless to yellow
100.0
(4-Chlorophenyl)acetonitrile
ClC6H4CH2CN
4-Chlorobenzyl cyanide
98+%
Glass bottle
1.5415 to 1.5435 (20°C, 589nm)
≥98.0%
250g
09, 448
98.0
140-53-4
MFCD00001918
98+%
C8H6ClN
0.3% max. (K.F.)
Authentic

Chemical Identifiers

140-53-4
100.0
C8H6ClN
151.6
(4-Chlorophenyl)acetonitrile
≥98.0%
98.0
98+%
ClC6H4CH2CN
MFCD00001918
4-Chlorobenzyl cyanide, 98+%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
May cause respiratory irritation.
Toxic in contact with skin.
Toxic if inhaled.
Toxic if swallowed.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger

EINECSNumber : 205-418-9

RTECSNumber : AL8250000

TSCA : TSCA