Please login to your online account to display your discounted pricing

4-Chlorobenzyl cyanide, 98+%, ACROS Organics™


Quantity 100g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  108811000

 View more versions of this product

Catalog No. AC108811000

  • / Each
 

Description & Specifications

Chemical Identifiers

Physical Form Liquid After Melting
Assay ≥98.0%
Formula Weight 151.6
Quantity 100g
Color Clear colorless to yellow
Beilstein 09, 448
EINECS Number 205-418-9
TSCA TSCA
RTECS Number AL8250000
CAS Max % 100.0
CAS Min % 98.0
Chemical Name or Material 4-Chlorobenzyl cyanide
Water 0.3% max. (K.F.)
Name Note 98+%
Assay Percent Range 98+%
Molecular Formula C8H6ClN
Linear Formula ClC6H4CH2CN
MDL Number MFCD00001918
CAS 140-53-4
Synonym (4-Chlorophenyl)acetonitrile
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.5415 to 1.5435 (20°C, 589nm)
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Toxic in contact with skin.
Toxic if inhaled.
Toxic if swallowed.
Causes serious eye irritation.
Causes skin irritation.

Specifications

Chemical Name or Material 4-Chlorobenzyl cyanide
Name Note 98+%
CAS 140-53-4
CAS Min % 98.0
CAS Max % 100.0
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C8H6ClN
Linear Formula ClC6H4CH2CN
Formula Weight 151.6
MDL Number MFCD00001918
Synonym (4-Chlorophenyl)acetonitrile
Accessories

Danger

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Toxic in contact with skin.
Toxic if inhaled.
Toxic if swallowed.
Causes serious eye irritation.
Causes skin irritation.

SDS & Safety Documents