missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Chlorobenzyl Alcohol 99%, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 108770250

 View more versions of this product

Catalog No. AC108770250


 

Specifications Description & Specifications

Specifications

142.58
≥98.5%
Almost white to beige
06, 444
100.0
ClC6H4CH2OH
4-Chlorobenzyl alcohol
C7H7ClO
99%
Authentic
Crystalline Needles or Powder
25g
07,59
69° to 72°C
99%
MFCD00004652
98.5
873-76-7
Glass bottle

Chemical Identifiers

873-76-7
100.0
C7H7ClO
142.58
4-Chlorobenzyl alcohol, 99%
98.5
99%
ClC6H4CH2OH
MFCD00004652
≥98.5%

Safety and Handling

SDS & Safety Documents

EINECSNumber : 212-852-2

RTECSNumber : DN9242000