Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chlorobenzoylacetonitrile, 98%

$91.73 - $1,112.91

Chemical Identifiers

Chemical Name or Material 4-Chlorobenzoylacetonitrile
CAS 4640-66-8
Molecular Formula C9H6ClNO
Formula Weight 179.61
MDL Number MFCD00051625
Beilstein 743368
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1236406

 
alfa aesar™
A1236406
5g Each for $91.73

AAA1236422

 
alfa aesar™
A1236422
100g Each for $1,112.91
Description & Specifications

Specifications

Formula Weight 179.61
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 4-Chlorobenzoylacetonitrile
Assay 98%
Molecular Formula C9H6ClNO
MDL Number MFCD00051625
CAS 4640-66-8
Beilstein 743368
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 128° to 132°C
Quantity 5g

Applications
Pharmaceutical Intermediates.

Solubility
Soluble in Dichloromethane, Ether, Ethyl Acetate and Methanol

Notes
Store at -20°C. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.