Learn More
4-Chlorobenzoyl chloride, 99+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 150191000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-Chlorobenzoyl chloride | |
12.0°C to 14.0°C | |
1.3700g/mL | |
105°C | |
99% min. (GC) | |
C7H4Cl2O | |
ClC6H4COCl | |
09,341 | |
benzoyl chloride, 4-chloro, p-chlorobenzoyl chloride, para-chlorobenzoyl chloride, 4-chlorobenzoic acid chloride, 4-chlorobenzoylchloride, 4-chloro benzoyl chloride, benzoylchloride, 4-chloro, benzoyl chloride, p-chloro, ccris 8935, pcboc | |
RKIDDEGICSMIJA-UHFFFAOYSA-N | |
4-chlorobenzoyl chloride | |
8501 | |
175.01 | |
Liquid |
122-01-0 | |
Colorless | |
222.0°C | |
Authentic | |
Glass bottle | |
1.5770 to 1.5790 | |
100 mL | |
1.37 | |
Solubility in water: hydrolyses. Other solubilities: soluble in alcohol,ether and acetone | |
C1=CC(=CC=C1C(=O)Cl)Cl | |
175.01 | |
CHEBI:60716 | |
99+% |
Chemical Identifiers
122-01-0 | |
175.01 | |
benzoyl chloride, 4-chloro, p-chlorobenzoyl chloride, para-chlorobenzoyl chloride, 4-chlorobenzoic acid chloride, 4-chlorobenzoylchloride, 4-chloro benzoyl chloride, benzoylchloride, 4-chloro, benzoyl chloride, p-chloro, ccris 8935, pcboc | |
CHEBI:60716 | |
C1=CC(=CC=C1C(=O)Cl)Cl |
C7H4Cl2O | |
RKIDDEGICSMIJA-UHFFFAOYSA-N | |
8501 | |
4-chlorobenzoyl chloride |
Safety and Handling
GHS H Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat
GHS Signal Word: Danger
EINECSNumber : 204-515-3
RUO – Research Use Only