missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Chlorobenzotrifluoride, 98%, Thermo Scientific™

Manufacturer:  Thermo Fisher Scientific AC108730010

 View more versions of this product

Catalog No. AC108730010

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Specifications

4-Chlorobenzotrifluoride
47°C
Glass bottle
1.35
136.0°C to 138.0°C
-36.0°C
98%
97.5
98%
ClC6H4CF3
15, 2127
QULYNCCPRWKEMF-UHFFFAOYSA-N
1-chloro-4-(trifluoromethyl)benzene
7394
Liquid
1.3500g/mL
Authentic
1.4453 to 1.4473
0.67 mPa.s (38°C)
Colorless
1L
98-56-6
100.0
C7H4ClF3
MFCD00000627
1-chloro-4-trifluoromethyl benzene, 4-chloro-alpha,alpha,alpha-trifluorotoluene, 4-chlorobenzotrifluoride, benzene, 1-chloro-4-trifluoromethyl, p-chloro-a,a,a-trifluorotoluene, p-chlorobenzotrifluoride, p-chlorophenyl trifluoromethane, p-chlorotrifluoromethylbenzene, para-chlorobenzotrifluoride, pcbtf
C1=CC(=CC=C1C(F)(F)F)Cl
180.56
180.56
98%

Chemical Identifiers

98-56-6
180.56
QULYNCCPRWKEMF-UHFFFAOYSA-N
7394
C1=CC(=CC=C1C(F)(F)F)Cl
C7H4ClF3
MFCD00000627
1-chloro-4-trifluoromethyl benzene, 4-chloro-alpha,alpha,alpha-trifluorotoluene, 4-chlorobenzotrifluoride, benzene, 1-chloro-4-trifluoromethyl, p-chloro-a,a,a-trifluorotoluene, p-chlorobenzotrifluoride, p-chlorophenyl trifluoromethane, p-chlorotrifluoromethylbenzene, para-chlorobenzotrifluoride, pcbtf
1-chloro-4-(trifluoromethyl)benzene
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Toxic to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wash face,hands and any exposed skin thorou

GHS Signal Word: Danger

EINECSNumber : 202-681-1

RTECSNumber : XS9145000

TSCA : TSCA

SDS Documents
Product Certifications

RUO – Research Use Only