missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chlorobenzotrifluoride, 98+%

Manufacturer:  Alfa Aesar™ A1515422

 View more versions of this product

Catalog No. AAA1515422



Description

Description

Applications
4-Chlorobenzotrifluoride acts as an important intermediate for dyes. It is used as an active component of polyurethane finishes. It is also used in adhesives, pigments, sealant chemicals, paints and coatings. Further, it is used as an environmental friendly solvent for cleaning or degreasing and dielectric fluid. It has a high capacity to dissolve inks, which is used in the printing industry. In addition to this, it is also used as xylene replacement in cleaners, thinners and other aromatic hydrocarbon blends.

Solubility
Miscible with chloroform. Slightly miscible with water.

Notes
Store in a cool place. Light sensitive. Incompatible with sodium dimethyl sulfonate, strong bases and strong oxidizing agents.
Specifications

Specifications

4-Chlorobenzotrifluoride
136°C to 138°C
100g
C7H4ClF3
UN2234
14,2126
QULYNCCPRWKEMF-UHFFFAOYSA-N
1-chloro-4-(trifluoromethyl)benzene
7394
≥98%
47°C (116°F)
-36°C
98-56-6
MFCD00000627
510203
4-chlorobenzotrifluoride, p-chlorobenzotrifluoride, 1-chloro-4-trifluoromethyl benzene, benzene, 1-chloro-4-trifluoromethyl, 4-chloro-alpha,alpha,alpha-trifluorotoluene, para-chlorobenzotrifluoride, p-chlorophenyl trifluoromethane, p-chloro-a,a,a-trifluorotoluene, pcbtf, p-chlorotrifluoromethylbenzene
C1=CC(=CC=C1C(F)(F)F)Cl
180.554
180.56

Chemical Identifiers

98-56-6
180.554
QULYNCCPRWKEMF-UHFFFAOYSA-N
7394
C1=CC(=CC=C1C(F)(F)F)Cl
C7H4ClF3
MFCD00000627
4-chlorobenzotrifluoride, p-chlorobenzotrifluoride, 1-chloro-4-trifluoromethyl benzene, benzene, 1-chloro-4-trifluoromethyl, 4-chloro-alpha,alpha,alpha-trifluorotoluene, para-chlorobenzotrifluoride, p-chlorophenyl trifluoromethane, p-chloro-a,a,a-trifluorotoluene, pcbtf, p-chlorotrifluoromethylbenzene
1-chloro-4-(trifluoromethyl)benzene
Safety and Handling

Safety and Handling

GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: CHLOROBENZOTRIFLUORIDES

RTECSNumber : XS9145000

TSCA : TSCA

Documents