Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chlorobenzotrichloride, 98+%

Chemical Identifiers

Chemical Name or Material 4-Chlorobenzotrichloride
CAS 5216-25-1
Molecular Formula C7H4Cl4
Formula Weight 229.92
MDL Number MFCD00000790
UN Number UN1760
Beilstein 1866549
Synonym alpha,alpha,alpha,4-Tetrachlorotoluene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2344614

  SDS
alfa aesar™
B2344614
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2344636

  SDS
alfa aesar™
B2344636
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2344622

  SDS
alfa aesar™
B2344622
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 229.92
Chemical Name or Material 4-Chlorobenzotrichloride
TSCA TSCA
Assay ≥98%
Molecular Formula C7H4Cl4
MDL Number MFCD00000790
CAS 5216-25-1
UN Number UN1760
Beilstein 1866549
Synonym alpha,alpha,alpha,4-Tetrachlorotoluene
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H350-H372-H361-H302-H312-H315-H335
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P260-P261-P280-P304+P340-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.
RTECS Number XT8580000
Flash Point 140°C (284°F)
Boiling Point 245°C
Melting Point 5° to 6°C
Quantity 25g
Safety and Handling

Danger

GHS P Statement
P260-P261-P280-P304+P340-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H350-H372-H361-H302-H312-H315-H335
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.