Learn More
4-Chlorobenzotrichloride, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2344614
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-Chlorobenzotrichloride | |
5216-25-1 | |
245°C | |
1.573 | |
MFCD00000790 | |
1866549 | |
LVZPKYYPPLUECL-UHFFFAOYSA-N | |
1-chloro-4-(trichloromethyl)benzene | |
21277 | |
≥98% |
5°C to 6°C | |
1.495 | |
25 g | |
C7H4Cl4 | |
UN1760 | |
4-chlorobenzotrichloride, 1-chloro-4-trichloromethyl benzene, p-chlorobenzotrichloride, benzene, 1-chloro-4-trichloromethyl, p-chlorophenyltrichloromethane, p-trichloromethylchlorobenzene, alpha,alpha,alpha,4-tetrachlorotoluene, unii-1up9gju33s, ccris 8902, p,alpha,alpha,alpha-tetrachlorotoluene | |
C1=CC(=CC=C1C(Cl)(Cl)Cl)Cl | |
229.909 | |
229.92 |
Chemical Identifiers
5216-25-1 | |
229.909 | |
LVZPKYYPPLUECL-UHFFFAOYSA-N | |
21277 | |
C1=CC(=CC=C1C(Cl)(Cl)Cl)Cl |
C7H4Cl4 | |
MFCD00000790 | |
4-chlorobenzotrichloride, 1-chloro-4-trichloromethyl benzene, p-chlorobenzotrichloride, benzene, 1-chloro-4-trichloromethyl, p-chlorophenyltrichloromethane, p-trichloromethylchlorobenzene, alpha,alpha,alpha,4-tetrachlorotoluene, unii-1up9gju33s, ccris 8902, p,alpha,alpha,alpha-tetrachlorotoluene | |
1-chloro-4-(trichloromethyl)benzene |
Safety and Handling
GHS H Statement
H350-H372-H361-H302-H312-H315-H335
May cause cancer.
Causes damage to organs through prolonged or repeated exposure.
Suspected of damaging fertility or the unborn child.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
May cause respiratory irritation.
P201-P202-P260-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c
H302+H312-H315-H319-H335-H350-H361f-H372
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.
EINECSNumber : 226-009-1
RTECSNumber : XT8580000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only