Please login to your online account to display your discounted pricing

4-Chlorobenzophenone 99%, ACROS Organics™


Quantity 500g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  108695000

 View more versions of this product

Catalog No. AC108695000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Powder
Formula Weight 216.67
Quantity 500g
Color White to off-white
Beilstein 07, 419
EINECS Number 205-160-7
TSCA TSCA
Melting Point 75° to 77°C
CAS Max % 100.0
Chemical Name or Material 4-Chlorobenzophenone
Name Note 99%
Molecular Formula C13H9ClO
MDL Number MFCD00000622
CAS 134-85-0
Infrared Spectrum Authentic
Assay Percent Range 99%
Linear Formula ClC6H4COC6H5
CAS Min % 98.5
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Loss on Drying 0.5% max.
Packaging Plastic bottle

Specifications

Chemical Name or Material 4-Chlorobenzophenone
Name Note 99%
CAS 134-85-0
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C13H9ClO
Linear Formula ClC6H4COC6H5
Formula Weight 216.67
MDL Number MFCD00000622
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents