Please login to your online account to display your discounted pricing

4-Chlorobenzophenone 99%, ACROS Organics™


Quantity 100g
Packaging Plastic bottle

Manufacturer: fisher scientific acros  108691000

 View more versions of this product

Catalog No. AC108691000

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Formula Weight 216.67
Physical Form Powder
Quantity 100g
Color White to off-white
Beilstein 07, 419
TSCA TSCA
Melting Point 75° to 77°C
EINECS Number 205-160-7
CAS Max % 100.0
Name Note 99%
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Loss on Drying 0.5% max.
Linear Formula ClC6H4COC6H5
CAS 134-85-0
Molecular Formula C13H9ClO
MDL Number MFCD00000622
Chemical Name or Material 4-Chlorobenzophenone
Assay Percent Range 99%
CAS Min % 98.5
Packaging Plastic bottle
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 4-Chlorobenzophenone
Name Note 99%
CAS 134-85-0
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C13H9ClO
Linear Formula ClC6H4COC6H5
Formula Weight 216.67
MDL Number MFCD00000622
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

SDS & Safety Documents