missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chlorobenzonitrile, 99%

Manufacturer:  Alfa Aesar™ A1452430

 View more versions of this product

Catalog No. AAA1452430



Description

Description

Applications
4-Chlorobenzonitrile is used to prepare a red pigment for plastics. It is also used in the preparation of amides in organic synthesis. Further, it is used as an intermediate of dye and active pharmaceutical ingredients.

Solubility
Soluble in alcohol and ether. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
Specifications

Specifications

4-Chlorobenzonitrile
Almond-like
90°C to 94°C
623-03-0
MFCD00001813
p-chlorobenzonitrile, benzonitrile, 4-chloro, p-cyanochlorobenzene, benzonitrile, p-chloro, 4-cyanochlorobenzene, nitril kyseliny p-chlorbenzoove, p-chlorbenzonitrile, unii-4z0hgp3a8a, pcbn, 4-chloro benzonitrile
C1=CC(=CC=C1C#N)Cl
137.566
137.57
108°C (226°F)
222°C to 224°C
250g
C7H4ClN
1072122
GJNGXPDXRVXSEH-UHFFFAOYSA-N
4-chlorobenzonitrile
12163
99%

Chemical Identifiers

623-03-0
137.566
GJNGXPDXRVXSEH-UHFFFAOYSA-N
12163
C1=CC(=CC=C1C#N)Cl
C7H4ClN
MFCD00001813
p-chlorobenzonitrile, benzonitrile, 4-chloro, p-cyanochlorobenzene, benzonitrile, p-chloro, 4-cyanochlorobenzene, nitril kyseliny p-chlorbenzoove, p-chlorbenzonitrile, unii-4z0hgp3a8a, pcbn, 4-chloro benzonitrile
4-chlorobenzonitrile
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H319
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.

GHS P Statement
P280h-P273-P305+P351+P338
Wear protective gloves/clothing.
Avoid release to the environment.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

RTECSNumber : DI2800000

TSCA : TSCA

Documents