Learn More
4-Chlorobenzonitrile, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1452418
Description
4-Chlorobenzonitrile is used to prepare a red pigment for plastics. It is also used in the preparation of amides in organic synthesis. Further, it is used as an intermediate of dye and active pharmaceutical ingredients.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Chlorobenzonitrile is used to prepare a red pigment for plastics. It is also used in the preparation of amides in organic synthesis. Further, it is used as an intermediate of dye and active pharmaceutical ingredients.
Solubility
Soluble in alcohol and ether. Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
4-Chlorobenzonitrile | |
623-03-0 | |
222°C to 224°C | |
Almond-like | |
MFCD00001813 | |
p-chlorobenzonitrile, benzonitrile, 4-chloro, p-cyanochlorobenzene, benzonitrile, p-chloro, 4-cyanochlorobenzene, nitril kyseliny p-chlorbenzoove, p-chlorbenzonitrile, unii-4z0hgp3a8a, pcbn, 4-chloro benzonitrile | |
GJNGXPDXRVXSEH-UHFFFAOYSA-N | |
4-chlorobenzonitrile | |
12163 | |
99% |
90°C to 94°C | |
1.2 | |
50 g | |
C7H4ClN | |
1072122 | |
Soluble in alcohol and ether. Insoluble in water. | |
ClC1=CC=C(C=C1)C#N | |
137.57 | |
137.57 |
Chemical Identifiers
623-03-0 | |
137.57 | |
GJNGXPDXRVXSEH-UHFFFAOYSA-N | |
12163 | |
ClC1=CC=C(C=C1)C#N |
C7H4ClN | |
MFCD00001813 | |
p-chlorobenzonitrile, benzonitrile, 4-chloro, p-cyanochlorobenzene, benzonitrile, p-chloro, 4-cyanochlorobenzene, nitril kyseliny p-chlorbenzoove, p-chlorbenzonitrile, unii-4z0hgp3a8a, pcbn, 4-chloro benzonitrile | |
4-chlorobenzonitrile |
Safety and Handling
GHS H Statement
H302-H312-H319
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye irritation.
P264b-P270-P280-P301+P312-P302+P352-P305+P351+P338-P312-P330-P363-P501c
H302+H312-H319
EINECSNumber : 210-765-4
RTECSNumber : DI2800000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only