missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chlorobenzoic acid, 98+%

Manufacturer:  Alfa Aesar™ A1513530

 View more versions of this product

Catalog No. AAA1513530



Description

Description

Applications
4-Chlorobenzoic acid is used to determine the diversity of 2-, 3- and 4-chlorobenzoate degraders in two pristine soils with similar physical and chemical characteristics.

Solubility
Soluble in water(0.08g/L).

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

4-Chlorobenzoic acid
Odorless
239°C to 243°C
74-11-3
MFCD00002531
14,2125
XRHGYUZYPHTUJZ-UHFFFAOYSA-N
4-chlorobenzoic acid
6318
156.57
238°C (460°F)
275°C
250g
C7H5ClO2
907196
p-chlorobenzoic acid, benzoic acid, 4-chloro, p-chlorbenzoic acid, chlorodracylic acid, benzoic acid, p-chloro, para-chlorobenzoic acid, 4-chloro-benzoic acid, p-carboxychlorobenzene, acido p-clorobenzoico, acido p-clorobenzoico italian
C1=CC(=CC=C1C(=O)O)Cl
156.565
CHEBI:30747
≥98%

Chemical Identifiers

74-11-3
156.565
XRHGYUZYPHTUJZ-UHFFFAOYSA-N
6318
4-chlorobenzoic acid
C7H5ClO2
MFCD00002531
p-chlorobenzoic acid, benzoic acid, 4-chloro, p-chlorbenzoic acid, chlorodracylic acid, benzoic acid, p-chloro, para-chlorobenzoic acid, 4-chloro-benzoic acid, p-carboxychlorobenzene, acido p-clorobenzoico, acido p-clorobenzoico italian
CHEBI:30747
C1=CC(=CC=C1C(=O)O)Cl
Safety and Handling

Safety and Handling

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280h-P305+P351+P338
Wear protective gloves/clothing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

RTECSNumber : DG4976010

TSCA : TSCA

Documents