missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-chlorobenzo[b]thiophene-2-carboxylic acid, 97%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC437960050
Specifications
4-chlorobenzo[b]thiophene-2-carboxylic acid | |
White | |
5g | |
96.0 | |
96% min. (HPLC) | |
2-carboxy-4-chlorobenzo b thiophene, 4-chloro benzo b thiophene-2-carboxylic acid, 4-chloro-2-benzo b thiophenecarboxylic acid, 4-chloro-benzo b thiophene-2-carboxylic acid, 4-chlorobenzo b thiophene-2-carboxylic acid, 4-chlorobenzothiophene-2-carboxylate, 4-chlorobenzothiophene-2-carboxylic acid, benzo b thiophene-2-carboxylic acid, 4-chloro | |
C1=CC2=C(C=C(S2)C(=O)O)C(=C1)Cl | |
212.66 | |
212.66 | |
97% |
Authentic | |
255°C to 256.7°C | |
23967-57-9 | |
100.0 | |
C9H5ClO2S | |
IPAXPERGAMNMIJ-UHFFFAOYSA-N | |
4-chloro-1-benzothiophene-2-carboxylic acid | |
3854516 | |
Crystalline Powder |
Chemical Identifiers
23967-57-9 | |
212.66 | |
2-carboxy-4-chlorobenzo b thiophene, 4-chloro benzo b thiophene-2-carboxylic acid, 4-chloro-2-benzo b thiophenecarboxylic acid, 4-chloro-benzo b thiophene-2-carboxylic acid, 4-chlorobenzo b thiophene-2-carboxylic acid, 4-chlorobenzothiophene-2-carboxylate, 4-chlorobenzothiophene-2-carboxylic acid, benzo b thiophene-2-carboxylic acid, 4-chloro | |
4-chloro-1-benzothiophene-2-carboxylic acid |
C9H5ClO2S | |
IPAXPERGAMNMIJ-UHFFFAOYSA-N | |
3854516 | |
C1=CC2=C(C=C(S2)C(=O)O)C(=C1)Cl |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortab
GHS Signal Word: Warning