Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chlorobenzil, 98+%

$21.99 - $415.92

Chemical Identifiers

Chemical Name or Material 4-Chlorobenzil
CAS 22711-23-5
Molecular Formula C14H9ClO2
Formula Weight 244.68g/mol
MDL Number MFCD00013646
Synonym 4-Chlorodibenzoyl
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1308106

 
alfa aesar™
A13081-06
5g Each for $109.99

AAA1308103

 
alfa aesar™
A13081-03
1g Each for $21.99

AAA1308114

 
alfa aesar™
A13081-14
25g Each for $415.92
Description & Specifications

Specifications

Melting Point 66° to 67°C
Quantity 5g
Chemical Name or Material 4-Chlorobenzil
CAS 22711-23-5
Assay ≥98%
Molecular Formula C14H9ClO2
Formula Weight 244.68g/mol
MDL Number MFCD00013646
Synonym 4-Chlorodibenzoyl