Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chlorobenzenesulfonyl chloride, 97%

Chemical Identifiers

Chemical Name or Material 4-Chlorobenzenesulfonyl chloride
CAS 98-60-2
Molecular Formula C6H4Cl2O2S
Formula Weight 211.07
MDL Number MFCD00007439
UN Number UN3261
Beilstein 511583
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1027122

  SDS
alfa aesar™
A1027122
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1027136

  SDS
alfa aesar™
A1027136
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 211.07
Health Hazard 2 GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Quantity 100g
Assay 97%
CAS 98-60-2
Melting Point 49° to 54°C
MDL Number MFCD00007439
Molecular Formula C6H4Cl2O2S
Health Hazard 3 GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 4-Chlorobenzenesulfonyl chloride
Health Hazard 1 Danger
RTECS Number DB8925000
Flash Point 107°C (224°F)
UN Number UN3261
Boiling Point 141°C (15mmHg)
DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA TSCA
Beilstein 511583
Safety and Handling

Danger

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.