Please login to your online account to display your discounted pricing

4-Chlorobenzenesulfonic Acid (Tech.), 90%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 4-Chlorobenzenesulfonic acid
Name Note tech., 90%
CAS 98-66-8
CAS Min % 88.0
CAS Max % 100.0
Purity Grade Technical
Assay 88% min.
Assay Percent Range 90%
Molecular Formula C6H5ClO3S
Linear Formula SO3HC6H4Cl
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC404500250

  SDS
thermo fisher scientific
404500250
Plastic bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Additional Information sulfones: < 2.0%
Hydrocarbons 0.5% max.
Infrared Spectrum Authentic
Packaging Plastic bottle
Sulfuric Acid sulfuric acid: < 3.5%
Water 3% max.
Color Gray-beige
Quantity 25g
Chemical Name or Material 4-Chlorobenzenesulfonic acid
Name Note tech., 90%
CAS 98-66-8
CAS Min % 88.0
CAS Max % 100.0
Purity Grade Technical
Physical Form Grayish-beige flaked solid
Assay 88% min.
Assay Percent Range 90%
Molecular Formula C6H5ClO3S
Linear Formula SO3HC6H4Cl
Formula Weight 192.62g/mol
MDL Number MFCD00065342
Merck Index 15, 2123
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
RTECS Number DB5074000
EINECS Number 202-690-0
TSCA TSCA
Safety and Handling

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.