Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chlorobenzenesulfonamide, 98%

$156.28 - $156.28

Chemical Identifiers

Chemical Name or Material 4-Chlorobenzenesulfonamide
CAS 98-64-6
Molecular Formula C6H6ClNO2S
Formula Weight 191.64g/mol
MDL Number MFCD00007936
Beilstein 1308891
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1673536

 
alfa aesar™
A16735-36
500g Each for $156.28

AAA1673522

 
alfa aesar™
A16735-22
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 144° to 148°C
Quantity 500g
Chemical Name or Material 4-Chlorobenzenesulfonamide
CAS 98-64-6
Assay 98%
Molecular Formula C6H6ClNO2S
Formula Weight 191.64g/mol
MDL Number MFCD00007936
Beilstein 1308891
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
RTECS Number DB1400000
TSCA TSCA
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.