Please login to your online account to display your discounted pricing

4-Chlorobenzenesulfinic acid sodium salt hydrate, 98%, Acros Organics

Chemical Identifiers

Chemical Name or Material 4-Chlorobenzenesulfinic acid sodium salt hydrate
CAS 14752-66-0
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5% (on dry substance)
Assay Percent Range 98%
Molecular Formula C6H4ClNaO2S·xH2O
Formula Weight 198.6
MDL Number MFCD00035602
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC355250010

  SDS
thermo fisher scientific
35525-0010
CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5% (on dry substance)
Physical Form Crystalline Powder
Formula Weight 198.6
Quantity 1g
Color White
EINECS Number 238-811-9
CAS Max % 100.0
Chemical Name or Material 4-Chlorobenzenesulfinic acid sodium salt hydrate
Water 14% max. (K.F.)
Assay Percent Range 98%
CAS Min % 97.5
Infrared Spectrum Authentic
Molecular Formula C6H4ClNaO2S·xH2O
MDL Number MFCD00035602
CAS 14752-66-0