Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chlorobenzeneseleninic acid, 99%

Chemical Identifiers

Chemical Name or Material 4-Chlorobenzeneseleninic acid
CAS 20753-53-1
Molecular Formula C6H5ClO2Se
Formula Weight 223.52
MDL Number MFCD00009981
UN Number UN3283
Synonym 4-Chlorophenylseleninic acid
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2118603

  SDS
alfa aesar™
B2118603
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2118606

  SDS
alfa aesar™
B2118606
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 223.52
MDL Number MFCD00009981
UN Number UN3283
Health Hazard 3 GHS P Statement
P273-P270-P309-P310-P302+P352-P501a
Avoid release to the environment.
Do not eat, drink or smoke when using this product.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material 4-Chlorobenzeneseleninic acid
Odor Unpleasant
Assay 99%
Molecular Formula C6H5ClO2Se
CAS 20753-53-1
Synonym 4-Chlorophenylseleninic acid
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H331-H373
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: SELENIUM COMPOUND, SOLID, N.O.S.
Melting Point 183° to 186°C
Quantity 1g
Safety and Handling

Danger

GHS P Statement
P273-P270-P309-P310-P302+P352-P501a
Avoid release to the environment.
Do not eat, drink or smoke when using this product.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H331-H373
Toxic if swallowed.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.