missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chlorobenzeneboronic acid pinacol ester, 97%

Manufacturer:  Alfa Aesar™ H5598403

 View more versions of this product

Catalog No. AAH5598403



Specifications

Specifications

4-Chlorobenzeneboronic acid pinacol ester
50°C to 55°C
195062-61-4
MFCD05663875
NYARTXMDWRAVIX-UHFFFAOYSA-N
2-(4-chlorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
10633712
Solid
>110°C (230°F)
Brown to White
1g
C12H16BClO2
1,3,2-dioxaborolane, 2-4-chlorophenyl-4,4,5,5-tetramethyl
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)Cl
238.518
238.52
97%

Chemical Identifiers

195062-61-4
238.518
NYARTXMDWRAVIX-UHFFFAOYSA-N
10633712
B1(OC(C(O1)(C)C)(C)C)C2=CC=C(C=C2)Cl
C12H16BClO2
MFCD05663875
1,3,2-dioxaborolane, 2-4-chlorophenyl-4,4,5,5-tetramethyl
2-(4-chlorophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Safety and Handling

Safety and Handling

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents