Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chlorobenzeneboronic acid pinacol ester, 97%

Chemical Identifiers

Chemical Name or Material 4-Chlorobenzeneboronic acid pinacol ester
CAS 195062-61-4
Formula Weight 238.52
MDL Number MFCD05663875
Synonym 4-Chlorophenylboronic acid pinacol ester; 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)chlorobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH5598403

 
alfa aesar™
H5598403
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5598406

 
alfa aesar™
H5598406
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH5598414

 
alfa aesar™
H5598414
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Melting Point 50° to 55°C
Formula Weight 238.52
Quantity 1g
MDL Number MFCD05663875
Physical Form Solid
Assay 97%
Color White to pale brown
Chemical Name or Material 4-Chlorobenzeneboronic acid pinacol ester
Synonym 4-Chlorophenylboronic acid pinacol ester; 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)chlorobenzene
CAS 195062-61-4