Please login to your online account to display your discounted pricing

4-Chlorobenzamidine hydrochloride, 95%, Acros Organics

Chemical Identifiers

Chemical Name or Material 4-Chlorobenzamidine hydrochloride
Assay Percent Range 95%
Molecular Formula C7H7ClN2·ClH
Formula Weight 191.06
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC448660500

 
thermo fisher scientific
AC448660500
50g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC448660050

  SDS
thermo fisher scientific
AC448660050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 191.06
Quantity 50g
Packaging Glass bottle
Assay Percent Range 95%
Chemical Name or Material 4-Chlorobenzamidine hydrochloride
CAS 14401-51-5
Molecular Formula C7H7ClN2·ClH