missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Chlorobenzamidine hydrochloride, 95%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific AC448660500
Specifications
4-Chlorobenzamidine hydrochloride | |
244.0°C to 245.0°C | |
14401-51-5 | |
C7H7ClN2·ClH | |
4-chloro-benzamidine hydrochloride, 4-chlorobenzamidine hcl, 4-chlorobenzamidine hydrochloride, 4-chlorobenzene-1-carboximidamide hydrochloride, 4-chlorobenzenecarboximidamide monohydrochloride, 4-chlorobenzimidamide hydrochloride, benzamide, p-chloro-, hydrochloride 7ci, benzamidine, p-chloro-, monohydrochloride, benzenecarboximidamide, 4-chloro-, monohydrochloride, p-chlorobenzamidine hydrochloride | |
C1=CC(=CC=C1C(=N)N)Cl.Cl | |
191.06 | |
191.06 |
Glass bottle | |
50g | |
94 to max. 106% min. (Argentometry) | |
ClC6H4C(=NH)NH2·HCl | |
RXAOGVQDNBYURA-UHFFFAOYSA-N | |
4-chlorobenzenecarboximidamide;hydrochloride | |
203436 | |
95% |
Chemical Identifiers
14401-51-5 | |
191.06 | |
4-chloro-benzamidine hydrochloride, 4-chlorobenzamidine hcl, 4-chlorobenzamidine hydrochloride, 4-chlorobenzene-1-carboximidamide hydrochloride, 4-chlorobenzenecarboximidamide monohydrochloride, 4-chlorobenzimidamide hydrochloride, benzamide, p-chloro-, hydrochloride 7ci, benzamidine, p-chloro-, monohydrochloride, benzenecarboximidamide, 4-chloro-, monohydrochloride, p-chlorobenzamidine hydrochloride | |
4-chlorobenzenecarboximidamide;hydrochloride |
C7H7ClN2·ClH | |
RXAOGVQDNBYURA-UHFFFAOYSA-N | |
203436 | |
C1=CC(=CC=C1C(=N)N)Cl.Cl |