missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chlorobenzamidine hydriodide, 96%

Manufacturer:  Alfa Aesar™ A1157106

 View more versions of this product

Catalog No. AAA1157106


 

Specifications

Specifications

4-Chlorobenzamidine hydriodide
5g
96%
MFCD00051987
YMEVGWRZEYGNJO-UHFFFAOYSA-N
4-chlorobenzenecarboximidamide;hydroiodide
2734221
~235°C (decomposition)
115297-57-9
C7H8ClIN2
8280383
C1=CC(=CC=C1C(=N)N)Cl.I
282.509
282.51

Chemical Identifiers

115297-57-9
282.509
YMEVGWRZEYGNJO-UHFFFAOYSA-N
C7H8ClIN2
MFCD00051987
2734221
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

SDS Documents