Learn More
4-Chlorobenzamide, 98+%, ACROS Organics™
Chemical Identifiers
CAS | 619-56-7 |
---|---|
Molecular Formula | C7H6ClNO |
Molecular Weight (g/mol) | 155.581 |
MDL Number | MFCD00007993 |
InChI Key | BLNVISNJTIRAHF-UHFFFAOYSA-N |
Synonym | 4-chloranylbenzamide, 4-chloro-benzamide, 4-chlorobenzenecarboxamide, benzamide, 4-chloro, benzamide, p-chloro, benzamide, p-chloro-6ci,7ci,8ci, p-chlorobenzamide, p-chlorobenzoic acid amide, pubchem3619, unii-260cv6k0mr |
PubChem CID | 12084 |
IUPAC Name | 4-chlorobenzamide |
SMILES | C1=CC(=CC=C1C(=O)N)Cl |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC108620100
|
Thermo Fisher Scientific
108620100 |
10g | Glass Bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
619-56-7 | |
155.581 | |
BLNVISNJTIRAHF-UHFFFAOYSA-N | |
12084 | |
C1=CC(=CC=C1C(=O)N)Cl |
C7H6ClNO | |
MFCD00007993 | |
4-chloranylbenzamide, 4-chloro-benzamide, 4-chlorobenzenecarboxamide, benzamide, 4-chloro, benzamide, p-chloro, benzamide, p-chloro-6ci,7ci,8ci, p-chlorobenzamide, p-chlorobenzoic acid amide, pubchem3619, unii-260cv6k0mr | |
4-chlorobenzamide |
Specifications
Authentic | |
White | |
10g | |
98.0 | |
98+% | |
ClC6H4CONH2 | |
09, 341 | |
BLNVISNJTIRAHF-UHFFFAOYSA-N | |
4-chlorobenzamide | |
12084 | |
Fine Crystalline Powder | |
4-Chlorobenzamide, 98+% |
Glass Bottle | |
177.0°C to 181.0°C | |
619-56-7 | |
100.0 | |
C7H6ClNO | |
MFCD00007993 | |
4-chloranylbenzamide, 4-chloro-benzamide, 4-chlorobenzenecarboxamide, benzamide, 4-chloro, benzamide, p-chloro, benzamide, p-chloro-6ci,7ci,8ci, p-chlorobenzamide, p-chlorobenzoic acid amide, pubchem3619, unii-260cv6k0mr | |
C1=CC(=CC=C1C(=O)N)Cl | |
155.581 | |
155.58 | |
≥98% |
Safety and Handling
GHS H Statement:
Harmful if swallowed.
Harmful if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber : 210-601-1
RTECSNumber : CV2443811