missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
4-Chloroanisole, 99%, ACROS Organics™
Chemical Identifiers
CAS | 623-12-1 |
---|---|
Molecular Formula | C7H7ClO |
Molecular Weight (g/mol) | 142.58 |
InChI Key | YRGAYAGBVIXNAQ-UHFFFAOYSA-N |
Synonym | 1-chloro-4-methoxy-benzene, 4-chloroanisole, 4-chlorophenol methyl ether, anisole, p-chloro, anisyl chloride, benzene, 1-chloro-4-methoxy, p-chloroanisole, p-chloromethoxybenzene, p-chlorophenyl methyl ether, unii-0f18bly08q |
PubChem CID | 12167 |
IUPAC Name | 1-chloro-4-methoxybenzene |
SMILES | COC1=CC=C(C=C1)Cl |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC170151000
|
Thermo Fisher Scientific
AC170151000 |
100g | Glass bottle |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
623-12-1 | |
142.58 | |
1-chloro-4-methoxy-benzene, 4-chloroanisole, 4-chlorophenol methyl ether, anisole, p-chloro, anisyl chloride, benzene, 1-chloro-4-methoxy, p-chloroanisole, p-chloromethoxybenzene, p-chlorophenyl methyl ether, unii-0f18bly08q | |
1-chloro-4-methoxybenzene |
C7H7ClO | |
YRGAYAGBVIXNAQ-UHFFFAOYSA-N | |
12167 | |
COC1=CC=C(C=C1)Cl |
Specifications
1.1600g/mL | |
Authentic | |
1.5340 to 1.5360 | |
198.0°C to 202.0°C | |
-18.0°C | |
623-12-1 | |
100.0 | |
C7H7ClO | |
06,186 | |
Solubility in water: immiscible. | |
COC1=CC=C(C=C1)Cl | |
142.58 | |
142.58 | |
99% |
78°C | |
Glass bottle | |
1.16 | |
Colorless to Yellow | |
100g | |
98.5 | |
98.5% min. (GC) | |
ClC6H4OCH3 | |
1-chloro-4-methoxy-benzene, 4-chloroanisole, 4-chlorophenol methyl ether, anisole, p-chloro, anisyl chloride, benzene, 1-chloro-4-methoxy, p-chloroanisole, p-chloromethoxybenzene, p-chlorophenyl methyl ether, unii-0f18bly08q | |
YRGAYAGBVIXNAQ-UHFFFAOYSA-N | |
1-chloro-4-methoxybenzene | |
12167 | |
Liquid | |
4-Chloroanisole, 99% |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and kee
GHS Signal Word: Warning
EINECSNumber : 210-772-2
TSCA : TSCA