missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloroaniline hydrochloride, 97%

Manufacturer:  Alfa Aesar™ L1374414

 View more versions of this product

Catalog No. AAL1374414



Specifications

Specifications

4-Chloroaniline hydrochloride
>230°C (sublimation)
20265-96-7
MFCD00042057
3560964
ISJBQSJDQZLCSF-UHFFFAOYSA-N
4-chloroaniline;hydrochloride
5284363
97%
Characteristic
25g
C6H7Cl2N
UN2018
4-chloroaniline hydrochloride, p-chloroaniline hydrochloride, 4-chlorobenzenamine hydrochloride, 4-chloroanilinium chloride, benzenamine, 4-chloro-, hydrochloride, p-chloroanilinium chloride, unii-po3w01cfsw, p-chloroaniline hcl, 4-chloroaniline hcl, p-chlorophenylamine hydrochloride
C1=CC(=CC=C1N)Cl.Cl
164.029
164.03

Chemical Identifiers

20265-96-7
164.029
ISJBQSJDQZLCSF-UHFFFAOYSA-N
5284363
C1=CC(=CC=C1N)Cl.Cl
C6H7Cl2N
MFCD00042057
4-chloroaniline hydrochloride, p-chloroaniline hydrochloride, 4-chlorobenzenamine hydrochloride, 4-chloroanilinium chloride, benzenamine, 4-chloro-, hydrochloride, p-chloroanilinium chloride, unii-po3w01cfsw, p-chloroaniline hcl, 4-chloroaniline hcl, p-chlorophenylamine hydrochloride
4-chloroaniline;hydrochloride
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H331-H350-H317
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause cancer.
May cause an allergic skin reaction.

GHS P Statement
P210-P273-P309-P310-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No Smoking.
Avoid release to the environment.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: CHLOROANILINES, SOLID

RTECSNumber : BX1394000

TSCA : TSCA

Documents