Please login to your online account to display your discounted pricing

4-Chloroaniline, 98%, ACROS Organics™


Quantity 5kg
Packaging Plastic Bottle

Manufacturer: thermo fisher scientific  AC108590051

 View more versions of this product

Catalog No. AC108590051

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Crystalline Solid
Assay ≥97.5%
Formula Weight 127.57
Quantity 5kg
Color Beige to brown-purple
Packaging Plastic Bottle
RTECS Number BX0700000
TSCA TSCA
EINECS Number 203-401-0
Melting Point 68° to 72°C
CAS Max % 100.0
Health Hazard 2 GHS H Statement:
May cause cancer.
Toxic if inhaled.
Very toxic to aquatic life with long lasting effects.
May cause an allergic skin reaction.
Toxic in contact with skin.
Toxic if swallowed.
Health Hazard 1 GHS Signal Word: Danger
Assay Percent Range 98%
Linear Formula ClC6H4NH2
CAS 106-47-8
CAS Min % 97.5
Chemical Name or Material 4-Chloroaniline
MDL Number MFCD00007835
Name Note 98%
Molecular Formula C6H6ClN
Storage Information The product turns dark during prolonged storage
Infrared Spectrum Authentic
Synonym 4-Chlorobenzenamine
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 4-Chloroaniline
Name Note 98%
CAS 106-47-8
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C6H6ClN
Linear Formula ClC6H4NH2
Formula Weight 127.57
MDL Number MFCD00007835
Synonym 4-Chlorobenzenamine
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause cancer.
Toxic if inhaled.
Very toxic to aquatic life with long lasting effects.
May cause an allergic skin reaction.
Toxic in contact with skin.
Toxic if swallowed.

SDS & Safety Documents