missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Chloroaniline, 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 108595000

 View more versions of this product

Catalog No. AC108595000


 

Specifications Description & Specifications

Specifications

≥97.5%
Crystalline Solid
Beige to brown-purple
100.0
Authentic
106-47-8
C6H6ClN
98%
ClC6H4NH2
4-Chlorobenzenamine
127.57
500g
68° to 72°C
The product turns dark during prolonged storage
MFCD00007835
4-Chloroaniline
98%
97.5
Glass bottle

Chemical Identifiers

106-47-8
100.0
C6H6ClN
127.57
4-Chlorobenzenamine
≥97.5%
97.5
98%
ClC6H4NH2
MFCD00007835
4-Chloroaniline, 98%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
May cause cancer.
Toxic if inhaled.
Very toxic to aquatic life with long lasting effects.
May cause an allergic skin reaction.
Toxic in contact with skin.
Toxic if swallowed.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Avoid release to the environment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 203-401-0

RTECSNumber : BX0700000

TSCA : TSCA