missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4'-Chloroacetoacetanilide 98%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 158555000

 View more versions of this product

Catalog No. AC158555000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥97.5%
211.65
Off-white to beige
100.0
4'-Chloroacetoacetanilide
C10H10ClNO2
MFCD00000613
98%
Plastic bottle
Crystalline Powder
500g
129° to 133°C
98%
97.5
101-92-8
CH3COCH2CONHC6H4Cl
Authentic

Chemical Identifiers

101-92-8
100.0
C10H10ClNO2
211.65
4'-Chloroacetoacetanilide, 98%
97.5
98%
CH3COCH2CONHC6H4Cl
MFCD00000613
≥97.5%

Safety and Handling

SDS & Safety Documents

EINECSNumber : 202-989-6

RTECSNumber : AK4375000

TSCA : TSCA