Please login to your online account to display your discounted pricing

4'-Chloroacetoacetanilide 98%, ACROS Organics™


Quantity 100g
Packaging Plastic bottle

Manufacturer: thermo fisher scientific  158551000

 View more versions of this product

Catalog No. AC158551000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Formula Weight 211.65
Physical Form Crystalline Powder
Quantity 100g
Color Off-white to beige
TSCA TSCA
RTECS Number AK4375000
Melting Point 129° to 133°C
EINECS Number 202-989-6
CAS Max % 100.0
Packaging Plastic bottle
MDL Number MFCD00000613
CAS Min % 97.5
Name Note 98%
Chemical Name or Material 4'-Chloroacetoacetanilide
Molecular Formula C10H10ClNO2
Assay Percent Range 98%
CAS 101-92-8
Linear Formula CH3COCH2CONHC6H4Cl
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 4'-Chloroacetoacetanilide
Name Note 98%
CAS 101-92-8
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10H10ClNO2
Linear Formula CH3COCH2CONHC6H4Cl
Formula Weight 211.65
MDL Number MFCD00000613