missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4'-Chloroacetoacetanilide, 98%, ACROS Organics™

Chemical Identifiers

CAS 101-92-8
Molecular Formula C10H10ClNO2
Molecular Weight (g/mol) 211.65
MDL Number MFCD00000613
InChI Key JMRJWEJJUKUBEA-UHFFFAOYSA-N
Synonym 4'-chloroacetoacetanilide, acetoacet-p-chloroanilide, acetoacetanilide, 4'-chloro, acetoacetanilide, p-chloro, acetoacetyl-4-chloroanilide, butanamide, n-4-chlorophenyl-3-oxo, n-4-chlorophenyl acetoacetamide, n-4-chlorophenyl-3-oxobutanamide, p-chloroacetoacetanilide, p-chlorodiacetanilide
PubChem CID 7587
ChEBI CHEBI:35090
IUPAC Name N-(4-chlorophenyl)-3-oxobutanamide
SMILES CC(=O)CC(=O)NC1=CC=C(C=C1)Cl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC158550050
SDS View Documents
Thermo Fisher Scientific
158550050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

101-92-8
211.65
JMRJWEJJUKUBEA-UHFFFAOYSA-N
7587
N-(4-chlorophenyl)-3-oxobutanamide
C10H10ClNO2
MFCD00000613
4'-chloroacetoacetanilide, acetoacet-p-chloroanilide, acetoacetanilide, 4'-chloro, acetoacetanilide, p-chloro, acetoacetyl-4-chloroanilide, butanamide, n-4-chlorophenyl-3-oxo, n-4-chlorophenyl acetoacetamide, n-4-chlorophenyl-3-oxobutanamide, p-chloroacetoacetanilide, p-chlorodiacetanilide
CHEBI:35090
CC(=O)CC(=O)NC1=CC=C(C=C1)Cl

Specifications

Authentic
White to Beige
5g
97.5
98%
CH3COCH2CONHC6H4Cl
4'-chloroacetoacetanilide, acetoacet-p-chloroanilide, acetoacetanilide, 4'-chloro, acetoacetanilide, p-chloro, acetoacetyl-4-chloroanilide, butanamide, n-4-chlorophenyl-3-oxo, n-4-chlorophenyl acetoacetamide, n-4-chlorophenyl-3-oxobutanamide, p-chloroacetoacetanilide, p-chlorodiacetanilide
CC(=O)CC(=O)NC1=CC=C(C=C1)Cl
211.65
CHEBI:35090
Crystalline Powder
4'-Chloroacetoacetanilide, 98%
Glass bottle
129°C to 133°C
101-92-8
100.0
C10H10ClNO2
MFCD00000613
JMRJWEJJUKUBEA-UHFFFAOYSA-N
N-(4-chlorophenyl)-3-oxobutanamide
7587
211.65
98%
Safety and Handling

Safety and Handling

EINECSNumber : 202-989-6

RTECSNumber : AK4375000

TSCA : TSCA

SDS