Learn More
4'-Chloroacetanilide, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1121436
Description
4'-Chloroacetanilide is used in pharmaceutical preparations, using PGC column. And also used in the synthesis of alprazolam.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4′-Chloroacetanilide is used in pharmaceutical preparations, using PGC column. And also used in the synthesis of alprazolam.
Solubility
Insoluble in water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
4'-Chloroacetanilide | |
178°C to 180°C | |
333°C | |
MFCD00000612 | |
509638 | |
4'-chloroacetanilide, n-4-chlorophenyl acetamide, 4-chloroacetanilide, p-chloroacetanilide, n-acetyl-p-chloroaniline, acetamide, n-4-chlorophenyl, acetanilide, 4'-chloro, acetic-4-chloroanilide, n-p-chlorophenyl acetamide, n-4-chloro-phenyl-acetamide | |
GGUOCFNAWIODMF-UHFFFAOYSA-N | |
N-(4-chlorophenyl)acetamide | |
10871 | |
169.61 |
539-03-7 | |
1.385 | |
C8H8ClNO | |
500 g | |
14,2111 | |
Insoluble in water. | |
CC(=O)NC1=CC=C(C=C1)Cl | |
169.608 | |
CHEBI:116915 | |
≥98% |
Chemical Identifiers
539-03-7 | |
169.608 | |
GGUOCFNAWIODMF-UHFFFAOYSA-N | |
10871 | |
N-(4-chlorophenyl)acetamide |
C8H8ClNO | |
MFCD00000612 | |
4'-chloroacetanilide, n-4-chlorophenyl acetamide, 4-chloroacetanilide, p-chloroacetanilide, n-acetyl-p-chloroaniline, acetamide, n-4-chlorophenyl, acetanilide, 4'-chloro, acetic-4-chloroanilide, n-p-chlorophenyl acetamide, n-4-chloro-phenyl-acetamide | |
CHEBI:116915 | |
CC(=O)NC1=CC=C(C=C1)Cl |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
EINECSNumber : 208-707-8
RTECSNumber : AE1001000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only