missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

4-Chloro-o-xylene 98%, ACROS Organics™

Chemical Identifiers

CAS 615-60-1
Molecular Formula C8H9Cl
Molecular Weight (g/mol) 140.61
MDL Number MFCD00000596
InChI Key HNQLMBJUMVLFCF-UHFFFAOYSA-N
Synonym 4-chloro-o-xylene, 3,4-dimethylchlorobenzene, 1-chloro-3,4-dimethylbenzene, benzene, 4-chloro-1,2-dimethyl, o-xylene, 4-chloro, 4-chloro-ortho-xylene, 4-chloro-1,2-xylene, unii-qo24waw04q, qo24waw04q, pubchem3641
PubChem CID 32886
IUPAC Name 4-chloro-1,2-dimethylbenzene
SMILES CC1=C(C=C(C=C1)Cl)C
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC160130050 SDS View Documents Acros Organics
160130050
5mL Glass bottle N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

615-60-1
140.61
HNQLMBJUMVLFCF-UHFFFAOYSA-N
32886
CC1=C(C=C(C=C1)Cl)C
C8H9Cl
MFCD00000596
4-chloro-o-xylene, 3,4-dimethylchlorobenzene, 1-chloro-3,4-dimethylbenzene, benzene, 4-chloro-1,2-dimethyl, o-xylene, 4-chloro, 4-chloro-ortho-xylene, 4-chloro-1,2-xylene, unii-qo24waw04q, qo24waw04q, pubchem3641
4-chloro-1,2-dimethylbenzene

Specifications

1.0470g/mL
Authentic
1.5273 to 1.5293
Colorless
615-60-1
100.0
C8H9Cl
MFCD00000596
4-chloro-o-xylene, 3,4-dimethylchlorobenzene, 1-chloro-3,4-dimethylbenzene, benzene, 4-chloro-1,2-dimethyl, o-xylene, 4-chloro, 4-chloro-ortho-xylene, 4-chloro-1,2-xylene, unii-qo24waw04q, qo24waw04q, pubchem3641
CC1=C(C=C(C=C1)Cl)C
140.61
140.61
98%
67°C
Glass bottle
221.0°C to 223.0°C
5mL
97.5
98%
(CH3)2C6H3Cl
05, 363
HNQLMBJUMVLFCF-UHFFFAOYSA-N
4-chloro-1,2-dimethylbenzene
32886
Liquid
4-Chloro-o-xylene, 98%
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful in contact with skin.
Causes serious eye irritation.
Harmful if inhaled.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 210-438-6