Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-o-xylene, 98%

$50.78 - $198.89

Chemical Identifiers

Chemical Name or Material 4-Chloro-o-xylene
CAS 615-60-1
Molecular Formula C8H9Cl
Formula Weight 140.61
MDL Number MFCD00000596
Beilstein 2076514
Synonym 1-Chloro-3,4-dimethylbenzene; 3,4-Dimethylchlorobenzene
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1903214

  SDS
alfa aesar™
A1903214
25g Each for $50.78

AAA1903222

  SDS
alfa aesar™
A1903222
100g Each for $198.89
Description & Specifications

Specifications

Formula Weight 140.61
Beilstein 2076514
Health Hazard 1 Warning
Chemical Name or Material 4-Chloro-o-xylene
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Boiling Point 221° to 223°C
Quantity 25g
Flash Point 67°C (153°F)
CAS 615-60-1
MDL Number MFCD00000596
Molecular Formula C8H9Cl
Synonym 1-Chloro-3,4-dimethylbenzene; 3,4-Dimethylchlorobenzene
Assay 98%
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.