Please login to your online account to display your discounted pricing

4-Chloro-o-phenylenediamine, 97%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  331860250

 View more versions of this product

Catalog No. AC331860250

  • / Each
 

Description & Specifications

Chemical Identifiers

Chemical Name or Material 4-Chloro-o-phenylenediamine
Color White to off-white
Melting Point 70° to 75°C
Quantity 25g
Name Note 97%
CAS 95-83-0
CAS Min % 96.0
Health Hazard 1 GHS Signal Word: Danger
CAS Max % 100.0
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Harmful if inhaled.
Harmful if swallowed.
Suspected of causing cancer.
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
RTECS Number SS8850000
EINECS Number 202-456-8
Packaging Glass bottle
TSCA TSCA
Water 3% max. (K.F.)
Physical Form Solid
Assay ≥96.0% (GC)
Assay Percent Range 97%
Molecular Formula C6H7ClN2
Linear Formula ClC6H3(NH)
Formula Weight 142.59
MDL Number MFCD00011691
Synonym 4-Chloro-1, 2-diaminobenzene

Specifications

Chemical Name or Material 4-Chloro-o-phenylenediamine
Name Note 97%
CAS 95-83-0
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0% (GC)
Assay Percent Range 97%
Molecular Formula C6H7ClN2
Linear Formula ClC6H3(NH)
Formula Weight 142.59
MDL Number MFCD00011691
Synonym 4-Chloro-1, 2-diaminobenzene
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Harmful if inhaled.
Harmful if swallowed.
Suspected of causing cancer.

SDS & Safety Documents