Learn More
4-Chloro-o-phenylenediamine, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 331860050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
4-Chloro-o-phenylenediamine | |
97% | |
White | |
100.0 | |
Authentic | |
Solid | |
C6H7ClN2 | |
MFCD00011691 | |
4-chloro-1,2-diaminobenzene, 4-chloro-o-phenylenediamine, 4-chloro-1,2-phenylenediamine, 1,2-diamino-4-chlorobenzene, 1,2-benzenediamine, 4-chloro, ursol olive 6g, 3,4-diaminochlorobenzene, 4-chloro-1,2-benzenediamine, 2-amino-4-chloroaniline, p-chloro-o-phenylenediamine | |
BXIXXXYDDJVHDL-UHFFFAOYSA-N | |
4-chlorobenzene-1,2-diamine | |
7263 | |
142.59 |
70°C to 75°C | |
95-83-0 | |
96.0 | |
5 g | |
Glass bottle | |
96% min. (GC) | |
ClC6H3(NH2)2 | |
13,25 | |
Solubility in water: 16g/L (20°C). Other solubilities: soluble in diluted acids and dmf,soluble in benzene,ethanol,diethyl ether,,petroleum ether and toluene | |
NC1=CC=C(Cl)C=C1N | |
142.59 | |
CHEBI:82301 | |
97% |
Chemical Identifiers
95-83-0 | |
142.59 | |
BXIXXXYDDJVHDL-UHFFFAOYSA-N | |
7263 | |
4-chlorobenzene-1,2-diamine |
C6H7ClN2 | |
MFCD00011691 | |
4-chloro-1,2-diaminobenzene, 4-chloro-o-phenylenediamine, 4-chloro-1,2-phenylenediamine, 1,2-diamino-4-chlorobenzene, 1,2-benzenediamine, 4-chloro, ursol olive 6g, 3,4-diaminochlorobenzene, 4-chloro-1,2-benzenediamine, 2-amino-4-chloroaniline, p-chloro-o-phenylenediamine | |
CHEBI:82301 | |
NC1=CC=C(Cl)C=C1N |
Safety and Handling
GHS H Statement:
Causes serious eye damage.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Harmful if inhaled.
Harmful if swallowed.
Suspected of causing cancer.
GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Use personal protective equipment as required.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be c
GHS Signal Word: Danger
EINECSNumber : 202-456-8
RTECSNumber : SS8850000
TSCA : TSCA
RUO – Research Use Only