missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 4-Chloro-N-phenylbenzamide, 97%

Manufacturer:  Alfa Aesar™ H59590MD

 View more versions of this product

Catalog No. AAH59590MD



Specifications

Specifications

4-Chloro-N-phenylbenzamide
6833-15-4
MFCD00000620
SFHDVPIEJXCMBP-UHFFFAOYSA-N
4-chloro-N-phenylbenzamide
346669
Solid
250mg
C13H10ClNO
4-chlor-benzanilid
C1=CC=C(C=C1)NC(=O)C2=CC=C(C=C2)Cl
231.679
231.68
97%

Chemical Identifiers

6833-15-4
231.679
SFHDVPIEJXCMBP-UHFFFAOYSA-N
346669
C1=CC=C(C=C1)NC(=O)C2=CC=C(C=C2)Cl
C13H10ClNO
MFCD00000620
4-chlor-benzanilid
4-chloro-N-phenylbenzamide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

Documents